Proteintech
Manufacturer Code:152231AP150UL
Catalog # 50-173-0078
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NT5C2 Fusion Protein Ag7438 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5&prime nucleotidase cytosolic II cN II Cytosolic 5 nucleotidase II GMP NT5B NT5C2 NT5CP PNT5; 5'-nucleotidase (purine), cytosolic type B; 5'-nucleotidase, cytosolic II; Cytosolic 5'-nucleotidase II; Cytosolic purine 5'-nucleotidase; IMP-specific 5'-NT
Gene Aliases: 2010002I23Rik; cN-II; CnII; ENSMUSG00000025039; Gm9751; GMP; NT5B; NT5C2; NT5CP; PNT5; SPG45; SPG65
UniProt ID: (Human) P49902, (Mouse) Q3V1L4
Entrez Gene ID: (Human) 22978, (Mouse) 76952, (Rat) 100363253
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.