Abnova
Manufacturer Code:H00022978K
Catalog # 89-210-688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human NT5C2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human NT5C2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: 5'-nucleotidase (purine), cytosolic type B; Cytosolic 5'-nucleotidase II; Cytosolic purine 5'-nucleotidase; GMP NT5B PNT5 cN-II; IMP-specific 5'-NT
Gene Aliases: cN-II; GMP; NT5B; NT5C2; NT5CP; PNT5; SPG45; SPG65
UniProt ID: (Human) P49902
Entrez Gene ID: (Human) 22978
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.