Invitrogen
Manufacturer Code:PA563340
Catalog # PIPA563340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NT5C1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIRP CN-IB CN1B autoimmune infertility-related protein cytosolic 5&prime-nucleotidase 1B; Autoimmune infertility-related protein; cN-IB; cN1B; Cytosolic 5'-nucleotidase 1B; Cytosolic 5'-nucleotidase IB; testicular tissue protein Li 1
Gene Aliases: AIRP; CN-IB; CN1B; FKSG85; NT5C1B
UniProt ID: (Human) Q96P26
Entrez Gene ID: (Human) 93034
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.