Invitrogen
Manufacturer Code:PA523805
Catalog # PIPA523805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 3-32 amino acids from the N-terminal region of human NT5C1B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5'nucleotidase; AIRP CN-IB CN1B autoimmune infertility-related protein cytosolic 5&prime-nucleotidase 1B; Autoimmune infertility-related protein; cN-IB; cN1B; Cytosolic 5'-nucleotidase 1B; Cytosolic 5'-nucleotidase IB; testicular tissue protein Li 1
Gene Aliases: 4921514H13Rik; AIRP; CN-IB; CN1B; FKSG85; NT5C1B
UniProt ID: (Human) Q96P26, (Mouse) Q91YE9
Entrez Gene ID: (Human) 93034, (Mouse) 70881
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.