Abnova
Manufacturer Code:PAB17686
Catalog # 89-119-349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human NT5C1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of NT5C1B.
For Research Use Only
Protein Aliases: AIRP; Autoimmune infertility-related protein; CN-IB; CN1B; Cytosolic 5'-nucleotidase 1B; Cytosolic 5'-nucleotidase IB; MGC26640; testicular tissue protein Li 1
Gene Aliases: AIRP; CN-IB; CN1B; FKSG85; NT5C1B
UniProt ID: (Human) Q96P26
Entrez Gene ID: (Human) 93034
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.