Invitrogen
Manufacturer Code:PA554461
Catalog # PIPA554461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NT5C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5' nucleotidase, deoxy (pyrimidine), cytosolic type C; 5'(3')-deoxyribonucleotidase, cytosolic type; 5,3-nucleotidase, cytosolic; Cytosolic 5',3'-pyrimidine nucleotidase; Deoxy-5'-nucleotidase 1; DNT DNT1 P5N2 PN-I PN-II UMPH2 cdN dNT-1 5&prime nucleotidase deoxy (pyrimidine) cytosolic type C 5&prime(3&prime)-deoxyribonucleotidase cytosolic type cytosolic 5&prime3&prime-pyrimidine nucleotidase deoxy-5&prime-nucleotidase 1 uridine 5&prime-monophosphate phosphohydrolase 2 uridine 5-prime monophosphate hydrolase 2; dNT-1; epididymis luminal protein 74; uridine 5'-monophosphate phosphohydrolase 2; uridine 5-monophosphate phosphohydrolase 2; uridine 5-prime monophosphate hydrolase 2
Gene Aliases: cdN; DNT; dNT-1; DNT1; HEL74; NT5C; P5N2; PN-I; PN-II; UMPH2
UniProt ID: (Human) Q8TCD5
Entrez Gene ID: (Human) 30833, (Rat) 100360582
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.