Abnova
Manufacturer Code:H00030833K
Catalog # 89-202-329
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human NT5C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human NT5C peptide using ARM Technology.
For Research Use Only
Protein Aliases: 5' nucleotidase, deoxy (pyrimidine), cytosolic type C; 5'(3')-deoxyribonucleotidase, cytosolic type; Cytosolic 5',3'-pyrimidine nucleotidase; Deoxy-5'-nucleotidase 1; DNT DNT1 P5N2 PN-I PN-II UMPH2 cdN dNT-1; dNT-1; epididymis luminal protein 74; uridine 5'-monophosphate phosphohydrolase 2; uridine 5-prime monophosphate hydrolase 2
Gene Aliases: cdN; DNT; dNT-1; DNT1; HEL74; NT5C; P5N2; PN-I; PN-II; UMPH2
UniProt ID: (Human) Q8TCD5
Entrez Gene ID: (Human) 30833
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.