OriGene
Manufacturer Code:TA500034S
Catalog # 50-167-1343
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E8 |
Immunogen | Human recombit protein fragment corresponding to amino acids 81-210 human NT4 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: GLC10 GLC1O NT-4 NT-4/5 NT-5 NT4 NT5 NTF5; neurotrophic factor 4; neurotrophic factor 5; neurotrophin 5 (neurotrophin 4/5); Neurotrophin-4; Neurotrophin-5; Neutrophic factor 4; NT-4; NT-5
Gene Aliases: GLC10; GLC1O; NT-4; NT-4/5; NT-5; NT4; NT5; NTF4; NTF5
UniProt ID: (Human) P34130
Entrez Gene ID: (Human) 4909
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.