Invitrogen
Manufacturer Code:PA546042
Catalog # PIPA546042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human NSUN5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S rRNA (cytosine-C(5))-methyltransferase; NOL1-related protein; NOL1/NOP2/Sun domain family member 5; NOL1R; NOP2/Sun domain family member 5 FLJ10267 MGC986 NOL1 NOL1R NSUN5A WBSCR20 WBSCR20A p120; NOP2/Sun domain family, member 5; NOP2/Sun domain family, member 5A; putative methyltransferase NSUN5; Williams Beuren syndrome chromosome region 20A; Williams-Beuren syndrome chromosomal region 20A protein; Williams-Beuren syndrome critical region protein 20 copy A
Gene Aliases: NOL1; NOL1R; NSUN5; NSUN5A; p120; p120(NOL1); WBSCR20; WBSCR20A
UniProt ID: (Human) Q96P11
Entrez Gene ID: (Human) 55695
Molecular Function:
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.