Proteintech
Manufacturer Code:154491AP150UL
Catalog # 50-173-0237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NSUN5 Fusion Protein Ag7711 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 28S rRNA (cytosine-C(5))-methyltransferase; NOL1 NOL1 related protein NOL1R NSUN5 NSUN5A p120 WBSCR20 WBSCR20A Ynl022cL; NOL1-related protein; NOL1/NOP2/Sun domain family member 5; NOL1/NOP2/Sun domain family, member 5; NOL1R; NOP2/Sun domain family, member 5; NOP2/Sun domain family, member 5A; probable 28S rRNA (cytosine-C(5))-methyltransferase; putative methyltransferase NSUN5; Williams Beuren syndrome chromosome region 20A; Williams Beuren syndrome chromosome region 20A homolog; Williams-Beuren syndrome chromosomal region 20A protein; Williams-Beuren syndrome chromosomal region 20A protein homolog; Williams-Beuren syndrome critical region protein 20; Williams-Beuren syndrome critical region protein 20 copy A
Gene Aliases: 9830109N13Rik; AI326939; NOL1; NOL1R; NSUN5; NSUN5A; p120; p120(NOL1); RGD1309268; WBSCR20; WBSCR20A
UniProt ID: (Human) Q96P11, (Mouse) Q8K4F6
Entrez Gene ID: (Human) 55695, (Mouse) 100609, (Rat) 288595
Molecular Function:
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.