Novus Biologicals
Manufacturer Code:NBP219594
Catalog # NBP219594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human NSUN4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-methylcytosine rRNA methyltransferase NSUN4; EC 2.1.1 EC 2.1.1.- EC 2.1.1.29 MGC22960 NOL1/NOP2/Sun domain family 4 NOL1/NOP2/Sun domain family member 4 NOL1/NOP2/Sun domain family member 4 NOP2/Sun domain family member 4 putative methyltransferase NSUN4 RP4-603I14.2; NOL1/NOP2/Sun domain family member 4; NOP2/Sun domain family, member 4; putative methyltransferase NSUN4; sperm head and tail associated protein
Gene Aliases: NSUN4; SHTAP
UniProt ID: (Human) Q96CB9
Entrez Gene ID: (Human) 387338
Molecular Function: methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.