Invitrogen
Manufacturer Code:PA543243
Catalog # PIPA543243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NSUN3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NOL1/NOP2/Sun domain family 3; NOL1/NOP2/Sun domain family member 3; NOL1/NOP2/Sun domain family member 3-like; NOL1/NOP2/Sun domain family, member 3; NOP2/Sun domain family, member 3; NOP2/Sun RNA methyltransferase family member 3; putative methyltransferase NSUN3; tRNA (cytosine(34)-C(5))-methyltransferase, mitochondrial
Gene Aliases: 6720484A09Rik; AU022521; MST077; MSTP077; NSUN3; UG0651E06
UniProt ID: (Bovine) Q0P5D8, (Human) Q9H649, (Mouse) Q8CCT7
Entrez Gene ID: (Bovine) 533791, (Horse) 100072898, (Rabbit) 100343486, (Human) 63899, (Dog) 607320, (Mouse) 106338, (Rat) 100360437
Molecular Function: methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.