Invitrogen
Manufacturer Code:PA5115674
Catalog # PIPA5115674
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NSUN2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-methycytoisine methyltransferase; 5-methycytoisine methyltransferase D13Wsu123e hTrm4 Misu mRNA cytosine C(5)-methyltransferase MRT5 Myc-induced SUN domain containing protein myc-induced SUN domain-containing protein Myc-induced SUN-domain-containing protein NOL1/NOP2/Sun domain family 2 NOL1/NOP2/Sun domain family member 2 NOL1/NOP2/Sun domain family member 2 NOP2/Sun domain family member 2 NOP2/Sun RNA methyltransferase family member 2 NOP2/Sun RNA methyltransferase family member 2 Nsun2 RGD1311954 RNA cytosine C(5)-methyltransferase NSUN2 SAKI Substrate of AIM1/Aurora kinase B TRM4 tRNA (cytosine(34)-C(5))-methyltransferase tRNA (cytosine-5-)-methyltransferase NSUN2 tRNA cytosine C(5)-methyltransferase tRNA methyltransferase 4 homolog; hTrm4; Misu; mRNA cytosine C(5)-methyltransferase; Myc-induced SUN domain containing protein; Myc-induced SUN domain-containing protein; Myc-induced SUN-domain-containing protein; NOL1/NOP2/Sun domain family 2; NOL1/NOP2/Sun domain family member 2; NOL1/NOP2/Sun domain family, member 2; NOP2/Sun domain family, member 2; NOP2/Sun RNA methyltransferase family, member 2; RNA cytosine C(5)-methyltransferase NSUN2; Substrate of AIM1/Aurora kinase B; tRNA (cytosine(34)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase NSUN2; tRNA cytosine C(5)-methyltransferase; tRNA methyltransferase 4 homolog
Gene Aliases: D13Wsu123e; MISU; MRT5; NSUN2; RGD1311954; SAKI; TRM4
UniProt ID: (Human) B3KP09, (Mouse) Q1HFZ0
Entrez Gene ID: (Human) 54888, (Rat) 361191, (Mouse) 28114
Molecular Function:
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.