Invitrogen
Manufacturer Code:PA572714
Catalog # PIPA572714
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | CCDC55 antibody for a 16 amino acid synthetic peptide near the carboxy terminus of human CCDC55. The immunogen is within amino acids 430 - 480 of CCDC55. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: coiled-coil domain containing 55; Coiled-coil domain-containing protein 55; NSrp70; Nuclear speckle splicing regulatory protein 1; Nuclear speckle splicing regulatory protein 1 Coiled-coil domain-containing protein 55 Nuclear speckle-related protein 70 NSrp70 NSRP1 CCDC55 NSRP70; Nuclear speckle-related protein 70
Gene Aliases: 8030451K07; AI851076; AL022898; CCDC55; HSPC095; NSpr70; NSRP1; NSRP70; RGD1309863
UniProt ID: (Mouse) Q5NCR9, (Human) Q9H0G5, (Rat) Q4FZU3
Entrez Gene ID: (Mouse) 237859, (Human) 84081, (Rat) 303346
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.