Novus Biologicals
Manufacturer Code:NBP190548
Catalog # NBP190548
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KREVGVQSSERNQDRKESSPNSRAKDKFLDQERSNKMRNMAKDKERNQEKPSNSESSLGAKHRLTEEGQEKGKEQERPPEAVSKFAKRNNEETVM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCDC55 coiled-coil domain containing 55 coiled-coil domain-containing protein 55 DKFZp434K1421 FLJ37233 HSPC095 NSRP70 nuclear speckle splicing regulatory protein 1; coiled-coil domain containing 55; Coiled-coil domain-containing protein 55; NSrp70; Nuclear speckle splicing regulatory protein 1; Nuclear speckle-related protein 70
Gene Aliases: CCDC55; HSPC095; NSRP1; NSRP70
UniProt ID: (Human) Q9H0G5
Entrez Gene ID: (Human) 84081
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.