Novus Biologicals
Manufacturer Code:NBP294395100UL
Catalog # NB023340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human NSRP1 (NP_115517.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCDC55 coiled-coil domain containing 55 coiled-coil domain-containing protein 55 DKFZp434K1421 FLJ37233 HSPC095 NSRP70 nuclear speckle splicing regulatory protein 1; coiled-coil domain containing 55; Coiled-coil domain-containing protein 55; NSrp70; Nuclear speckle splicing regulatory protein 1; Nuclear speckle-related protein 70
Gene Aliases: CCDC55; HSPC095; NSRP1; NSRP70
UniProt ID: (Human) Q9H0G5
Entrez Gene ID: (Human) 84081
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.