Abnova
Manufacturer Code:MAB3340
Catalog # 89-111-360
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 31C4 |
Immunogen | Native purified NSR1 from yeast nuclear protein preparations. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native NSR1.
For Research Use Only
Protein Aliases: Nsr1p; Nuclear localization sequence-binding protein; p67; SHE5
Gene Aliases: G7001; NSR1; SHE5; YGR159C
UniProt ID: (Baker's yeast) P27476
Entrez Gene ID: (Baker's yeast) 853064
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.