Proteintech
Manufacturer Code:136271AP150UL
Catalog # 50-172-8888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NSMCE2 Fusion Protein Ag4545 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C8orf36 E3 SUMO protein ligase NSE2 hMMS21 MMS21 MMS21 homolog Non SMC element 2 homolog NSE2 NSMCE2; E3 SUMO-protein ligase NSE2; E3 SUMO-protein transferase NSE2; hMMS21; methyl methanesulfonate sensitivity gene 21; MMS21 homolog; Non-SMC element 2 homolog; non-SMC element 2, MMS21 homolog; Non-structural maintenance of chromosomes element 2 homolog; zinc finger, MIZ-type containing 7
Gene Aliases: 1110014D18Rik; AI661537; C8orf36; MMS21; NSE2; NSMCE2; RGD1305156; ZMIZ7
UniProt ID: (Human) Q96MF7, (Mouse) Q91VT1, (Rat) Q4V8A0
Entrez Gene ID: (Human) 286053, (Mouse) 68501, (Rat) 299957
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.