Invitrogen
Manufacturer Code:PA5101719
Catalog # PIPA5101719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NSG1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain neuron cytoplasmic protein 1; brain neuron cytoplasmic protein 1 brain neuron cytoplasmic protein 1/2 brain specific mRNA b Bsmrb carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa protein D4S234 D4S234E m234 NEEP21 neuron specific gene family member 1 Neuronal vesicle trafficking-associated protein 1 Neuron-enriched endosomal protein of 21 kDa Neuron-specific protein family member 1 Nsg1 p21 PEP1; Brain neuron cytoplasmic protein 1/2; brain specific mRNA b; carboxyterminally EE-tagged neuron-enriched endosomal 21 kDa protein; M234; Neuron-enriched endosomal protein of 21 kDa; Neuron-specific protein family member 1; Neuronal vesicle trafficking-associated protein 1; p21
Gene Aliases: Bsmrb; D4S234; D4S234E; m234; NEEP21; NSG1; P21; PEP1
UniProt ID: (Human) P42857, (Mouse) Q62092, (Rat) P02683
Entrez Gene ID: (Human) 27065, (Mouse) 18196, (Rat) 25247
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.