Invitrogen
Manufacturer Code:PA521633
Catalog # PIPA521633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 21 and 302 of NSFL1C (Uniprot IDNo.Q9UNZ2) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NSFL1 (p97) cofactor (p47); NSFL1 cofactor p47; p97 cofactor p47; RP4-776F14.2 P47 UBX1 UBXD10 UBXN2C dJ776F14.1 NSFL1 cofactor p47 SHP1 homolog UBX domain-containing protein 2C p97 cofactor p47; SHP1 homolog; UBX domain-containing protein 2C; XY body-associated protein XY40
Gene Aliases: dJ776F14.1; Munc-18c; NSFL1C; P47; Stxbp3a; UBX1; UBXD10; UBXN2C; XY40
UniProt ID: (Human) Q9UNZ2, (Rat) O35987, (Mouse) Q9CZ44
Entrez Gene ID: (Human) 55968, (Rat) 83809, (Mouse) 386649
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.