Abnova
Manufacturer Code:H00055968K
Catalog # 89-210-609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human NSFL1C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human NSFL1C peptide using ARM Technology.
For Research Use Only
Protein Aliases: MGC3347 UBX1 UBXD10 UBXN2C dJ776F14.1 p47; NSFL1 (p97) cofactor (p47); NSFL1 cofactor p47; p97 cofactor p47; SHP1 homolog; UBX domain-containing protein 2C
Gene Aliases: dJ776F14.1; NSFL1C; P47; UBX1; UBXD10; UBXN2C
UniProt ID: (Human) Q9UNZ2
Entrez Gene ID: (Human) 55968
Molecular Function: membrane traffic protein membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.