Abnova
Manufacturer Code:PAB9620
Catalog # 89-134-278
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus rat Nsf. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Nsf.
For Research Use Only
Protein Aliases: AI316878; AU020090; AU067812; N-ethylmaleimide sensitive factor; N-ethylmaleimide-sensitive fusion protein; NEM-sensitive fusion protein; Protein SKD2; SKD2; Suppressor of K(+) transport growth defect 2; Vesicle-fusing ATPase; Vesicular-fusion protein NSF
Gene Aliases: AI316878; AU020090; AU067812; Nsf; SKD2
UniProt ID: (Mouse) P46460
Entrez Gene ID: (Mouse) 18195
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.