Novus Biologicals
Manufacturer Code:NBP183306
Catalog # NBP183306
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:LPYAMKPIDYYTETKILQERAVLGANDPEKNFLTTAIRPHGIFGPRDPQLVPILIEAARNGKMKFVIGNGKNLVDFTFVENVVHGHILAAEQLSRDSTLGGKAFHITNDEPIPFWTFLSRI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.1.1.170 H105e3 member 1 NAD(P) dependent steroid dehydrogenase-like SDR31E1 sterol-4-alpha-carboxylate 3-dehydrogenase decarboxylating XAP104; Protein H105e3; short chain dehydrogenase/reductase family 31E, member 1; Sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating
Gene Aliases: H105E3; NSDHL; SDR31E1; XAP104
UniProt ID: (Human) Q15738
Entrez Gene ID: (Human) 50814
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.