Proteintech
Manufacturer Code:151111AP150UL
Catalog # 50-172-9988
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NSDHL Fusion Protein Ag7097 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bare patches; H105E3 NSDHL Protein H105e3 SDR31E1 XAP104; NAD(P)-dependent steroid dehydrogenase-like protein; Protein H105e3; short chain dehydrogenase/reductase family 31E, member 1; Sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating; striated
Gene Aliases: AI747449; Bpa; H105E3; NSDHL; SDR31E1; Str; XAP104
UniProt ID: (Human) Q15738, (Mouse) Q9R1J0, (Rat) Q5PPL3
Entrez Gene ID: (Human) 50814, (Mouse) 18194, (Rat) 309262
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.