Novus Biologicals
Manufacturer Code:NBP104324AF532
Catalog # NP104324A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7B4 |
Immunogen | Recombinant human NSD3 (383-660aa) purified from E. coli |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 2.1.1.43 FLJ20353 histone-lysine N-methyltransferase NSD3 MGC126766 MGC142029 NSD3DKFZp667H044 Nuclear SET domain-containing protein 3 pp14328 Protein whistle WHSC1-like 1 isoform 9 with methyltransferase activity to lysine WHSC1-like protein 1 Wolf-Hirschhorn syndrome candidate 1-like 1 Wolf-Hirschhorn syndrome candidate 1-like protein 1; Histone-lysine N-methyltransferase NSD3; nuclear receptor binding SET domain protein 3; Nuclear SET domain-containing protein 3; Protein whistle; WHSC1-like 1 isoform 9 with methyltransferase activity to lysine; WHSC1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like protein 1
Gene Aliases: DC28; NSD3; pp14328; WHISTLE; WHSC1L1
UniProt ID: (Human) Q9BZ95
Entrez Gene ID: (Human) 54904
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.