Invitrogen
Manufacturer Code:PA582203
Catalog # PIPA582203
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NSD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ20353 KMT3F WHISTLE WHSC1L1; Histone-lysine N-methyltransferase NSD3; nuclear receptor binding SET domain protein 3; Nuclear SET domain-containing protein 3; Protein whistle; WHSC1-like 1 isoform 9 with methyltransferase activity to lysine; WHSC1-like protein 1; Wolf-Hirschhorn syndrome candidate 1-like protein 1
Gene Aliases: DC28; NSD3; pp14328; WHISTLE; WHSC1L1
UniProt ID: (Human) Q9BZ95
Entrez Gene ID: (Human) 54904
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.