Invitrogen
Manufacturer Code:PA553385
Catalog # PIPA553385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NSD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; MMSET; MMSET NSD2 REIIBP TRX5 WHS IL5 promoter REII region-binding protein multiple myeloma SET domain containing protein type III nuclear SET domain-containing protein 2 probable histone-lysine N-methyltransferase NSD2 trithorax/ash1-related protein 5; multiple myeloma SET domain containing protein type III; Multiple myeloma SET domain-containing protein; nuclear receptor binding SET domain protein 2; Nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; trithorax/ash1-related protein 5; Wolf-Hirschhorn syndrome candidate 1 protein
Gene Aliases: KIAA1090; MMSET; NSD2; REIIBP; TRX5; WHS; WHSC1
UniProt ID: (Human) O96028
Entrez Gene ID: (Human) 7468
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.