Invitrogen
Manufacturer Code:PA540725
Catalog # PIPA540725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human WHSC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; MMSET; multiple myeloma SET domain containing protein type III; Multiple myeloma SET domain-containing protein; nuclear receptor binding SET domain protein 2; Nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; putative histone-lysine N-methyltransferase NSD2-like protein; trithorax/ash1-related protein 5; Wolf-Hirschhorn syndrome candidate 1; Wolf-Hirschhorn syndrome candidate 1 protein; Wolf-Hirschhorn syndrome candidate 1 protein homolog; Wolf-Hirschhorn syndrome candidate 1 WHS NSD2 TRX5 MMSET REIIBP
Gene Aliases: 5830445G22Rik; 9430010A17Rik; AW555663; C130020C13Rik; D030027O06Rik; D930023B08Rik; KIAA1090; mKIAA1090; MMSET; NSD2; REIIBP; RGD1565590; TRX5; WHS; WHSC1; Whsc1l
UniProt ID: (Human) O96028, (Mouse) Q8BVE8
Entrez Gene ID: (Dog) 479077, (Horse) 100051090, (Rabbit) 100347860, (Guinea pig) 100713660, (Human) 7468, (Rat) 680537, (Mouse) 107823
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.