Proteintech
Manufacturer Code:239551AP150UL
Catalog # 50-173-4176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NSBP1 Fusion Protein Ag20884 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: High mobility group nucleosome-binding domain-containing protein 5; high-mobility group nucleosome binding domain 5; HMGN5 NSBP1 nucleosomal binding protein 1 Nucleosome binding protein 1; nucleosomal binding protein 1; Nucleosome-binding protein 1
Gene Aliases: HMGN5; NBP-45; NSBP1
UniProt ID: (Human) P82970
Entrez Gene ID: (Human) 79366
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.