Invitrogen
Manufacturer Code:PA589854
Catalog # PIPA589854
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-260 of human NSA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hairy cell leukemia protein 1; L-name related protein; L-name-related protein 42; LNR42; NSA2 ribosome biogenesis homolog; Ribosome biogenesis protein NSA2 homolog; TGF beta-inducible nuclear protein; TGF beta-inducible nuclear protein 1; TGF-beta inducible nuclear protein; TGF-beta-inducible nuclear protein 1
Gene Aliases: 5730427N09Rik; CDK105; HCL-G1; HCLG1; HUSSY-29; HUSSY29; LNR42; NSA2; Nsa2p; TINP1; Yr29
UniProt ID: (Human) O95478, (Mouse) Q9CR47
Entrez Gene ID: (Human) 10412, (Rat) 171456, (Mouse) 59050
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.