Invitrogen
Manufacturer Code:PA583659
Catalog # PIPA583659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NSA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ94393 HCLG1 HUSSY-29 TINP1; Hairy cell leukemia protein 1; NSA2 ribosome biogenesis homolog; Ribosome biogenesis protein NSA2 homolog; TGF beta-inducible nuclear protein 1; TGF-beta inducible nuclear protein; TGF-beta-inducible nuclear protein 1
Gene Aliases: CDK105; HCL-G1; HCLG1; HUSSY-29; HUSSY29; NSA2; TINP1
UniProt ID: (Human) O95478
Entrez Gene ID: (Human) 10412
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.