Novus Biologicals
Manufacturer Code:NBP287943
Catalog # NB035565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C terminal region of human NRXN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp313P2036 FLJ35941 KIAA0578Neurexin I-alpha neurexin 1 neurexin I Neurexin I-beta neurexin-1-alpha neurexin-1-beta PTHSL2; neurexin I; Neurexin I-alpha; Neurexin I-beta; Neurexin-1; Neurexin-1-alpha; Neurexin-1-beta
Gene Aliases: Hs.22998; KIAA0578; NRXN1; PTHSL2; SCZD17
UniProt ID: (Human) Q9C081
Entrez Gene ID: (Human) 9378
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.