Invitrogen
Manufacturer Code:PA537003
Catalog # PIPA537003
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 42-87 of human NRL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neural retina-specific leucine zipper protein; Neural retina-specific leucine zipper protein D14S46E; neural retinal-specific leucine zipper; NRL
Gene Aliases: D14H14S46E; D14S46E; NRL; NRL-MAF; RP27
UniProt ID: (Human) P54845, (Mouse) P54846
Entrez Gene ID: (Human) 4901, (Rat) 290221, (Mouse) 18185
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.