Novus Biologicals
Manufacturer Code:NBP2822930.1ML
Catalog # NB030222
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human NRL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D14S46Eneural retina-specific leucine zipper protein neural retina leucine zipper NRL-MAF RP27neural retinal-specific leucine zipper; Neural retina-specific leucine zipper protein; neural retinal-specific leucine zipper; NRL
Gene Aliases: D14S46E; NRL; NRL-MAF; RP27
UniProt ID: (Human) P54845
Entrez Gene ID: (Human) 4901
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.