Proteintech
Manufacturer Code:177041AP150UL
Catalog # 50-173-1730
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NRIP2 Fusion Protein Ag11989 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neuronal interacting factor 1; Neuronal-interacting factor X 1; NRIP2; nuclear receptor interacting protein 2; nuclear receptor interacting protein 2-like; Nuclear receptor-interacting protein 2
Gene Aliases: AW491344; NIX1; NRIP2
UniProt ID: (Human) Q9BQI9, (Mouse) Q9JHR9
Entrez Gene ID: (Human) 83714, (Rat) 689619, (Mouse) 60345
Molecular Function:
aspartic protease
hydrolase
protease
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.