Novus Biologicals
Manufacturer Code:NBP130075V
Catalog # NBP130075V
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Maps to a region between residue 300 and 350 of human nuclear receptor interaction protein (IQ motif and WD repeats 1) using the numbering given in entry NP_001017977.1 |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200006M05Rik Androgen receptor complex-associated protein ARCAP DDB1 and CUL4 associated factor 6 DDB1- and CUL4-associated factor 6 FLJ23798 IQ motif and WD repeat-containing protein 1 IQ motif and WD repeats 1 IQWD1 MSTP055 NRIP Nuclear receptor interaction protein PC326; Androgen receptor complex-associated protein; ARCAP; DDB1- and CUL4-associated factor 6; IQ motif and WD repeat-containing protein 1; IQ motif and WD repeats 1; NRIP; Nuclear receptor interaction protein
Gene Aliases: 1200006M05Rik; ARCAP; DCAF6; IQWD1; MSTP055; NRIP; PC326
UniProt ID: (Human) Q58WW2
Entrez Gene ID: (Human) 55827
Molecular Function:
G-protein
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.