Novus Biologicals
Manufacturer Code:NB10056434
Catalog # NB10056434
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 800-850 of human NRIP was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1200006M05Rik Androgen receptor complex-associated protein ARCAP DDB1 and CUL4 associated factor 6 DDB1- and CUL4-associated factor 6 FLJ23798 IQ motif and WD repeat-containing protein 1 IQ motif and WD repeats 1 IQWD1 MSTP055 NRIP Nuclear receptor interaction protein PC326; Androgen receptor complex-associated protein; ARCAP; DDB1- and CUL4-associated factor 6; IQ motif and WD repeat-containing protein 1; IQ motif and WD repeats 1; NRIP; Nuclear receptor interaction protein
Gene Aliases: 1200006M05Rik; ARCAP; DCAF6; IQWD1; MSTP055; NRIP; PC326
UniProt ID: (Human) Q58WW2
Entrez Gene ID: (Human) 55827
Molecular Function: G-protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.