Novus Biologicals
Manufacturer Code:NBP290070AF647
Catalog # NB069502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human NRG4 (Q8WWG1 Pro2-Phe62). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp779N0541 DKFZp779N1944 heregulin 4 HRG4pro-NRG4 neuregulin 4 pro-neuregulin-4 membrane-bound isoform Pro-NRG4; heregulin 4; Neuregulin-4; NRG-4; Pro-neuregulin-4, membrane-bound isoform; Pro-NRG4
Gene Aliases: HRG4; NRG4
UniProt ID: (Human) Q8WWG1
Entrez Gene ID: (Human) 145957
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.