Abnova
Manufacturer Code:MAB21222
Catalog # 89-337-715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | NRG1/2752 |
Immunogen | Recombinant protein corresponding to amino acids 21-242 of human NRG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody.
Protein Aliases: Acetylcholine receptor-inducing activity; ARIA; ARIA GGF GGF2 HGL HRG HRG1 HRGA NDF SMDF; Breast cancer cell differentiation factor p45; Glial growth factor; glial growth factor 2; Heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); HRG; Neu differentiation factor; Neuregulin-1; Pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; sensory and motor neuron derived factor; Sensory and motor neuron-derived factor
Gene Aliases: ARIA; GGF; GGF2; HGL; HRG; HRG1; HRGA; MST131; MSTP131; NDF; NRG1; NRG1-IT2; SMDF
UniProt ID: (Human) A5YAK5
Entrez Gene ID: (Human) 3084
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.