Invitrogen
Manufacturer Code:PA578627
Catalog # PIPA578627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of mouse Neuregulin-1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholine receptor-inducing activity; ARIA; Glial growth factor; Heregulin; HRG; Neu differentiation factor; neuregulin 1 type III beta 3; Neuregulin-1; Nrg1 neuregulin 1; Pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; Sensory and motor neuron-derived factor; type I neuregulin 1; type III neuregulin 1
Gene Aliases: 6030402G23Rik; ARIA; D230005F13Rik; GGF; GGFII; Hgl; HRG; HRGalpha; NDF; Nrg1; SMDF
UniProt ID: (Rat) P43322
Entrez Gene ID: (Rat) 112400, (Mouse) 211323
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.