Invitrogen
Manufacturer Code:PA516626
Catalog # PIPA516626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A highly antigenic synthetic peptide from the EGF-like domain of human heregulin |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholine receptor-inducing activity; ARIA; Breast cancer cell differentiation factor p45; Glial growth factor; glial growth factor 2; glial growth factor heregulin alpha (45kD ERBB2 p185-activator) neu differentiation factor pro-NRG1 pro-neuregulin-1 membrane-bound isoform sensory and motor neuron derived factor; Heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); HRG; Neu differentiation factor; Neuregulin-1; Pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; sensory and motor neuron derived factor; Sensory and motor neuron-derived factor; type I neuregulin 1; type III neuregulin 1
Gene Aliases: 6030402G23Rik; ARIA; D230005F13Rik; GGF; GGF2; GGFII; HGL; HRG; HRG1; HRGA; HRGalpha; MST131; MSTP131; NDF; NRG1; NRG1-IT2; SMDF
UniProt ID: (Human) A5YAK5
Entrez Gene ID: (Human) 3084, (Mouse) 211323
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.