Invitrogen
Manufacturer Code:PA527882
Catalog # PIPA527882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 108 and 413 of Human NRF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HEBP1; NF-E2-related factor 2; NFE2-related factor 2; NRF2 HEBP1 NF-E2-related factor 2 NFE2-related factor 2 nuclear factor erythroid 2-related factor 2 nuclear factor erythroid-derived 2-like 2 nuclear factor erythroid derived 2 like 2; Nuclear factor erythroid 2-related factor 2; nuclear factor erythroid-derived 2-like 2; nuclear factor, erythroid 2-like 2; Nuclear factor, erythroid derived 2, like 2
Gene Aliases: AI194320; HEBP1; NFE2L2; NRF2
UniProt ID: (Human) Q16236, (Mouse) Q60795
Entrez Gene ID: (Human) 4780, (Mouse) 18024
Molecular Function: actin family cytoskeletal protein cytoskeletal protein hydrolase non-motor actin binding protein nucleic acid binding protease serine protease transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.