Invitrogen
Manufacturer Code:PA579763
Catalog # PIPA579763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence in the middle region of human NRF1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha palindromic-binding protein; Alpha-pal; NRF-1; Nuclear respiratory factor 1; Nuclear respiratory factor 1 NRF-1 Alpha palindromic-binding protein Alpha-pal NRF1; nuclear respiratory factor-1
Gene Aliases: ALPHA-PAL; C87038; D6Ertd415e; Nfe2l1_retired; NRF1
UniProt ID: (Human) Q16656, (Mouse) Q9WU00
Entrez Gene ID: (Human) 4899, (Rat) 312195, (Mouse) 18181
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.