Invitrogen
Manufacturer Code:PA541485
Catalog # PIPA541485
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human NRF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha palindromic-binding protein; Alpha-pal; NRF-1; Nuclear respiratory factor 1; Nuclear respiratory factor 1 ALPHA-PAL EWG; nuclear respiratory factor-1
Gene Aliases: ALPHA-PAL; Nfe2l1_retired; NRF1
UniProt ID: (Human) Q16656
Entrez Gene ID: (Human) 4899, (Rat) 312195
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.