Invitrogen
Manufacturer Code:PA540912
Catalog # PIPA540912
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human NRF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha palindromic-binding protein; Alpha-pal; Not really finished protein; nrf; NRF-1; nrl; Nuclear respiratory factor 1; Nuclear respiratory factor 1 ALPHA-PAL; nuclear respiratory factor-1
Gene Aliases: ALPHA-PAL; C87038; D6Ertd415e; Nfe2l1_retired; NRF1; SC:BZ3C13.1; SC:bZ3C13.2; ube2h
UniProt ID: (Human) Q16656, (Mouse) Q9WU00, (Zebrafish) Q90X44
Entrez Gene ID: (Dog) 482260, (Bovine) 509232, (Human) 4899, (Horse) 100056975, (Rabbit) 100339141, (Rat) 312195, (Guinea pig) 100720898, (Mouse) 18181, (Zebrafish) 64604
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.