Novus Biologicals
Manufacturer Code:NBP230609
Catalog # NBP230609
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: PNNTALWQKYLLFCQSQFSTFSISKIHSLYGKCLSTLSAVKDGSILSHPALPGTEEAMFALFLQQCHFLRQAGHSEKAISLFQAMVDFTFFKPDSVKDLPTKGQVEFFEPFWDSGEPRAGEKGARGWKAWMHQQERGGWV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C14orf102 chromosome 14 open reading frame 102 FLJ10008 FLJ14051 hypothetical protein LOC55051; Nuclear exosome regulator NRDE2; Protein NRDE2 homolog; UPF0614 protein C14orf102
Gene Aliases: C14orf102; NRDE2
UniProt ID: (Human) Q9H7Z3
Entrez Gene ID: (Human) 55051
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.