Invitrogen
Manufacturer Code:MA527673
Catalog # PIMA527673
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S364-51 |
Immunogen | Fusion protein amino acids 30-845 (extracellular domain) of mouse NrCAM. Rat: 96&percnt identity (795/822 amino acids identical). Human: 91&percnt identity (753/822 amino acids identical) &bsim50&percnt identity with Neurofascin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ankyrin-binding cell adhesion molecule NrCAM; hBravo; mBravo; neuron-glia-CAM-related cell adhesion molecule; Neuronal cell adhesion molecule; Neuronal cell adhesion molecule Bravo hBravo KIAA0343 MGC138845 MGC138846 Neuronal surface protein Bravo Ng CAM related Ng-CAM-related NgCAM related cell adhesion molecule NgCAM-related cell adhesion molecule Nr CAM Nr-CAM; Neuronal surface protein Bravo; Ng-CAM-related; NgCAM-related cell adhesion molecule; Nr-CAM; rBravo
Gene Aliases: Bravo; C030017F07Rik; C130076O07Rik; KIAA0343; mKIAA0343; NRCAM
UniProt ID: (Human) Q92823, (Mouse) Q810U4
Entrez Gene ID: (Human) 4897, (Mouse) 319504, (Rat) 497815
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.