OriGene
Manufacturer Code:CF803854
Catalog # 50-166-6703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C1 |
Immunogen | Full length human recombit protein of human NRBF2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Comodulator of PPAR and RXR; comodulator of PPAR and RXR 1; comodulator of PPAR and RXR 2; COPR COPR1 COPR2 NRBF-2; NRBF-2; nuclear receptor binding factor-2; Nuclear receptor-binding factor 2
Gene Aliases: COPR; COPR1; COPR2; NRBF-2; NRBF2
UniProt ID: (Human) Q96F24
Entrez Gene ID: (Human) 29982
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.