Invitrogen
Manufacturer Code:PA531539
Catalog # PIPA531539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 83 and 287 of Human NRBF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Comodulator of PPAR and RXR; comodulator of PPAR and RXR 1; comodulator of PPAR and RXR 2; COPR1 COPR2 NRBF-2 comodulator of PPAR and RXR nuclear receptor binding factor-2 nuclear receptor-binding factor 2; NRBF-2; nuclear receptor binding factor-2; Nuclear receptor-binding factor 2
Gene Aliases: COPR; COPR1; COPR2; NRBF-2; NRBF2
UniProt ID: (Human) Q96F24, (Mouse) Q8VCQ3
Entrez Gene ID: (Human) 29982, (Mouse) 641340
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.