Proteintech
Manufacturer Code:107241AP150UL
Catalog # 50-172-6542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NRAS Fusion Protein Ag1081 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALPS4 GTPase NRas HRAS1 N ras NRAS NRAS1 Transforming protein N Ras; GTPase NRas; N-ras protein part 4; neuroblastoma RAS viral (v-ras) oncogene homolog; Transforming protein N-Ras; v-ras neuroblastoma RAS viral oncogene homolog
Gene Aliases: ALPS4; AV095280; CMNS; HRAS1; N-ras; NCMS; NRAS; NRAS1; NS6
UniProt ID: (Human) P01111, (Rat) Q04970
Entrez Gene ID: (Human) 4893, (Mouse) 18176, (Rat) 24605
Molecular Function: G-protein enzyme modulator small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.